Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAPP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552089
Description
Immunogen sequence: GAGAVGALQL TYISKVSIAT PKQKPKTPFC FVINALSQRY FLQANDQKDM KDWVEALNQA SKITVPKGGG LPMTTEVLKS LAAPPALEKK PQVAYKTEII GGVVVHTPIS QNGGDGQEGS EPGSHTILRR SQSYIPTS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
PLEKHA2 binds specifically to phosphatidylinositol 3, 4- diphosphate (PtdIns3, 4P2), but not to other phosphoinositides. It may recruit other proteins to the plasma membrane.
Specifications
TAPP2 | |
Polyclonal | |
Unconjugated | |
PLEKHA2 | |
PH domain-containing family A member 2; pleckstrin homology domain containing A2; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 2; pleckstrin homology domain-containing family A member 2; pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2; PLEKHA2; tandem PH Domain containing protein-2; tandem PH domain-containing protein 2; TAPP2; TAPP-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59339 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HB19 | |
PLEKHA2 | |
Recombinant protein corresponding to Human TAPP2. Recombinant protein control fragment (Product #RP-102146). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction