Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAPT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565524 Supplier Invitrogen™ Supplier No. PA565524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLNVAFNSHNK SLLTIMMSNN FVEIKGSVFK KFEKNNLFQM SNSDIKERFT NYVLLLIVCL RNMEQFSWNP DHL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

This gene encodes a highly conserved, putative transmembrane protein. A mutation in the mouse ortholog of this gene results in homeotic, posterior-to-anterior transformations of the axial skeleton which are similar to the phenotype of mouse homeobox C8 gene mutants. This gene is proposed to function downstream of homeobox C8 to transduce extracellular patterning information during axial skeleton development. An alternatively spliced transcript variant encoding a substantially different isoform has been described, but its biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAPT1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAPT1
Gene Accession No. Q6NXT6
Gene Alias 4932414K18Rik; CMVFR; Cytomegalovirus partial fusion receptor; OCLSBG; TAPT1; transmembrane anterior posterior transformation 1; transmembrane anterior posterior transformation protein 1; Transmembrane anterior posterior transformation protein 1 homolog
Gene Symbols TAPT1
Host Species Rabbit
Immunogen Recombinant Human TAPT1. Recombinant protein control fragment (Product #RP-106196).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202018
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.