Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Taq Core Kit 10

Catalog No. 11EPTQK105C Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 250U
1000U
5 x 1000U
5 x 250U
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11EPTQK105C 5 x 250U
11EPTQK300 1000U
11EPTQK109 10 x 250U
11EPTQK10L 5 x 1000U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11EPTQK105C Supplier MP Biomedicals™ Supplier No. 11EPTQK105C

Includes

Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix

Supplied with standard reaction buffers

Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.

Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix. Store at -20°C
Form Frozen
Format Frozen liquid
Quantity 5 x 250U
For Use With (Application) Standard PCR

Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.