Learn More
Description
Includes
Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix
Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.
Specifications
Specifications
| Content And Storage | Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix. Store at -20°C |
| Form | Frozen |
| Format | Frozen liquid |
| Quantity | 5 x 1000U |
| For Use With (Application) | Standard PCR |
Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.