Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Taq CORE Kit 25

Supplied with standard reaction buffers

Specifications

Content And Storage Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix. Store at -20°C
For Use With (Application) Standard PCR
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11EPTQK600
View Documents
MP Biomedicals™
11EPTQK600
12 x 600U N/A N/A N/A
11EPTQK259
View Documents
MP Biomedicals™
11EPTQK259
10 x 250U N/A N/A N/A
Description

Description

Specifications

Specifications

Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix. Store at -20°C
Standard PCR
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.