Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Taq DNA Polymerase (cloned)
GSA_VA

Catalog No. 46001008 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 x 250U
250U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-001-008 250U
46-001-010 10 x 250U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-001-008 Supplier Cytiva Supplier No. 27079804
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

For PCR and other procedures requiring DNA polymerase activity at elevated temperatures

One unit incorporates 10nmol of total nucleotides into acid-insoluble material in 30 min. at 70°C in a total volume of 50μL.

  • Purity: Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities
  • Concentration: 5000 units/mL
  • Functional testing: Tested in PCR
  • Storgae Buffer: 50mM Tris-HCl (pH 7.5), 5mM DTT, 0.1mM EDTA, 50% glycerol, stabilizers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5000 U/mL
Content And Storage 100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl. Store at -20°C
pH 7.5
Purity or Quality Grade Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities.
Quantity 250U
For Use With (Application) Standard PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.