Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TrueMark™ Comprehensive Microbiota Control

Catalog No. A50382
Encompass_Preferred
Change view
Click to view available options
Volume:
50 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume
A50382 1000 μL
A50383 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50382 Supplier Applied Biosystems™ Supplier No. A50382
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The TrueMark Comprehensive Microbiota Control contains a linearized multi-target plasmid pool with microbial target sequences commonly used on Custom OpenArray panels. The control has targets relevant to many microbial profiling applications, including respiratory tract, urinary tract, vaginal microbiota, and antibiotic resistance research.

The TrueMark Comprehensive Microbiota Control contains a linearized multi-target plasmid pool with microbial target sequences commonly used on Custom OpenArray panels. The control has targets relevant to many microbial profiling applications, including respiratory tract, urinary tract, vaginal microbiota, and antibiotic resistance research. The control can be included as a positive control for panel-specific amplification. It is available in two concentrations: 1 x 105 and 5 x 107 copies/μL. The higher concentration can be used to determine parameters necessary for analytical validation (e.g., limit of detection and Ct cut-off values).

NOTE: For Version 4 of TrueMark Comprehensive Microbiota Control, please contact your local representative.

The plasmid also contains target sequences for TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control, TaqMan Universal Extraction Control Organism (B. atrophaeus), and the human RNase P RPPH1 gene. During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and to help with troubleshooting.

Note: This product will be updated on an annual basis to ensure coverage of the most popular microbial targets, and performance will be measured to ensure consistency between versions.

Version history

Version 1 of this product was first available in April 2021 and contained 471 microbial assay targets.

Version 2 became available in October 2021 and contains an additional 83 microbial assay targets.

Version 3 became avalible in March 2023 and contains an additional 139 microbial and antibiotic resistant targets.

Version 4 became available in February 2025 and contains an additional 150 microbial and antibiotic resistant targets.

Version 5 became available in September 2026 and contains an additional 86 microbial and antibiotic resistant targets.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 x 105 Copies/μL
Content And Storage 1000 μL at a concentration of 1 x 105 copies/μL
Detection Method Primer-probe
Format Tube
Product Line TrueMark
Quantity 1000 μL (1 x 105 copies/μL)
Shipping Condition Dry ice
Volume 1000 μL
Which method/instrument was used to determine the concentration of the plasmid in the TrueMark Comprehensive Microbiota Control (Cat. No. A50382)?

The plasmid concentration was determined by means of digital PCR on the QuantStudio 3D Digital PCR System.

Are the targets in the TaqMan Comprehensive Microbiota Control (Cat. No. A50382) present as single copy?

Yes, they are. However, with the exception of Xeno and RNase P which are present as multiple copies.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.