Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan Fast Advanced Master Mix Gene Expression Bundle with MicroAmp Plates and MicroAmp Seals PROMO

Catalog No. A47377PM
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Real Time PCR (qPCR)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
A47377PM Real Time PCR (qPCR)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A47377PM Supplier Applied Biosystems™ Supplier No. A47377PM
Get Up to 48% Off Gene Expression Packages
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TaqMan Fast Advanced Master Mix delivers better sensitivity, greater specificity, and faster results.

  • Applied Biosystems has affordable qPCR tools that fit every budget and level of experience. Makes getting the best results from your data easy while saving money.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 5 mL Master Mix, 20 MicroAmp Plates, 25 MicroAmp Seals
For Use With (Application) Real Time PCR (qPCR)
Includes MicroAmp Plates, MicroAmp Seals
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.