Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Microbe Detection Assay, pooled

Catalog No. A52083
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A52083 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A52083 Supplier Applied Biosystems™ Supplier No. A52083
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applied Biosystems TaqMan Microbe Detection assays are used for quantitative real-time PCR detection of microbial species.

Applied Biosystems TaqMan Microbe Detection assays are used for quantitative real-time PCR detection of microbial species. These assays consist of up to four pooled unlabeled PCR primers and TaqMan probes with a dye label (FAM) on the 5’ end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3’ end.

Features of TaqMan Microbe Detection assays include:
• Easy to use—just add DNA and master mix and run the qPCR—no melt curves required
• Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
• Sensitive—TaqMan assays are ideal for measuring low-abundance microbial targets
• Accurate—identify microbial targets with high accuracy of quantitation
• Extensive content—over 300 pre-designed microbe detection assays currently available
• Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for closely related microbial species and subtypes
• Fast delivery—assays shipped in as little as 1–2 days (see below)

Approximate ship time
• Inventoried: 1–2 days in North America*
• Made to order: 4–6 days in North America

All of our predesigned TaqMan gene expression assays are covered by the TaqMan Assays qPCR Guarantee.**

*Our most popular assays are stocked in North America for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.

**Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

TRUSTED_SUSTAINABILITY

Specifications

3'Primer Modification FAM
Concentration 20X
5'Primer Modification MGB-NFQ
Content And Storage Store at -15 to -25°C.

Detection Method Primer-probe
Format Tube
PCR Method qPCR
Product Line TaqMan
Quantity 250 μL
Shipping Condition Dry Ice
For Use With (Application) Pathogen Detection
No. of Reactions 250 Reactions
Show More Show Less
What are the concentrations of the primers and probes for TaqMan microbial assays?

Our predesigned and custom TaqMan microbial 20X assays have a primer concentration of 900 nM and a probe concentration of 250 nM in the final 1X concentration.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.