Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan Oncology Multiplex Positive Control, 10% VAF

Catalog No. A40007974
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40007974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40007974 Supplier Applied Biosystems™ Supplier No. A40007974
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TaqMan Oncology Multiplex Positive Control, 10% VAF, is a specialized positive control reference material designed for liquid biopsy analysis of actionable cancer biomarkers.

The Applied Biosystems TaqMan Oncology Multiplex Positive Control, 10% VAF, is a specialized positive control reference material designed for cancer biomarkers for liquid biopsy research. This control material is carefully formulated to contain known cancer biomarkers at a standardized 10% variant allele frequency (VAF) across multiple genes, including BRAF, EGFR, KRAS, PIK3CA, and ESR1.

The product enables translational researchers and molecular pathologists to help ensure workflow integrity and help provide confidence in analytical research results.

Specifically designed for compatibility with TaqMan Oncology dPCR Multiplex kits, this positive control facilitates a turnkey solution when implemented on the QuantStudio Absolute Q dPCR System, offering a comprehensive quality control framework for oncology biomarker detection workflows.

Features

  • Ready-to-use format — carefully formulated positive control delivered in a ready-to-use configuration that enables a streamlined pipette-and-go workflow, reducing preparation steps and hands-on time
  • Wet lab verified performance — comprehensive wet laboratory testing conducted on the Absolute Q dPCR System to verify product characteristics and help ensure analytical reliability
  • Lot-to-lot reproducibility — manufactured using stringent Thermo Fisher Scientific manufacturing processes designed to deliver consistent and reproducible results across production lots

Why is this control formulated for liquid biopsy applications?

This positive control has been specifically formulated with liquid biopsy analysis requirements in mind, delivering a ready-to-use format that helps reduce the need for additional preparation steps. The control is engineered to accurately emulate the characteristics of fragmented DNA that would typically be extracted from appropriate plasma samples, to facilitate experimental conditions that closely mirror real-world clinical specimens. This design consideration is critical for liquid biopsy workflows, where the integrity and fragmentation patterns of circulating cell-free DNA directly impact assay performance and downstream analytical results.

How does the control integrate with biomarker assay workflows?

The implementation of this positive control follows a straightforward protocol that integrates easily with existing biomarker assay procedures. Users simply incorporate the positive control as instructed within their specific biomarker assay protocols, maintaining consistency with established laboratory workflows. The control is then pipetted directly into the MAP16 plate, which has been designed to accommodate the precise volumetric requirements of the assay system while helping to ensure optimal sample distribution and processing efficiency.

Is the control compatible with the Absolute Q system?

The positive control demonstrates compatibility with the Absolute Q platform, enabling direct execution of assays without requiring additional instrumentation or protocol modifications. Once the control has been properly loaded into the MAP16 plate according to the specified instructions, the entire assembly can be processed directly on the Absolute Q system. This easy integration helps support quality control measures that can be implemented without disrupting established analytical workflows or requiring specialized handling procedures that might introduce additional sources of variability into the experimental process.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 tube containing positive control for specific cancer biomarkers at 10% variant allele frequency. Store at 4°C
Detection Method Digital PCR
Form Liquid
Format Tube
GC-Rich PCR Performance Medium
PCR Method Digital PCR
Reaction Speed <2 hr.
For Use With Applied Biosystems TaqMan Oncology dPCR Multiplex Kits
Label or Dye Unlabeled
Product Type Positive Control
Purification Method Alkali based lysis followed by ethanol precipitation
Purity or Quality Grade Purified dsDNA fragments
Quantity 100 μL
Research Category RUO
Shipping Condition Frozen
For Use With (Application) Oncology, DNA Quantitation
Thermostability Up to 98°C
Final Product Type dPCR DNA positive control, fragmented to lengths similar to circulating blood DNA
Sample Type Synthetic
Starting Material Plasmid
Includes Control Template(s)
Storage Buffer Low EDTA Tris buffer pH 8.0 + tsRNA
Inhibitor Tolerance (Simple) Not inhibitory to PCR
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.