Learn More
Applied Biosystems™ TaqMan™ PreAmp Master Mix (cDNA pre-amplification for qPCR)
Description
- Analyze mRNA from precious samples such as laser capture microdissections, needle biopsies, and formalin-fixed paraffin-embedded tissues (FFPE)
- Stretch as little as 1ng of cDNA into 200 real-time PCR reactions for gene expression analysis using TaqMan™ Gene Expression assays
- Analyze up to 100 gene expression targets with minimal hands-on time
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
Specifications
| Content And Storage | Contains 2X TaqMan™ PreAmp Master Mix (1 mL). Store at 2–8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | AmpliTaq Gold DNA Polymerase |
| For Use With (Equipment) | 7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System |
| Passive Reference Dye | No ROX |
| Product Line | TaqMan |
| Product Type | PreAmp Master Mix |
| Quantity | 40 reactions |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.