Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control

Catalog No. 956126
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-612-6 10 x 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 95-612-6 Supplier Applied Biosystems™ Supplier No. 956126
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control contains targets specific to the SARS-CoV-2, influenza A/B, and RSV genomic regions targeted by the TaqMan SARS-CoV-2, FluA/B RSV RT-PCR Assay Kit.

The TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control is recommended for use with the TaqMan SARS-CoV-2, FluA/B RSV RT-PCR Assay Kit as a positive control to ensure reaction setup and reagent integrity when interpreting test results.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at ≤-70°C.
Format Tube(s)
For Use With TaqMan™ SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit
Product Line TaqMan
Quantity 10 x 10 μL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.