Learn More
Applied Biosystems™ TaqMan™ Universal RNA Spike In/Reverse Transcription (Xeno) Control
Description
The control is used with the proprietary TaqMan assay for Xeno sequences (Assay IDs Ac00010014_a1 or Pa00010014_a1). The TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control is supplied at a concentration of 10,000 copies/μL. During nucleic acid isolation, 10 μL of the control is added to each test sample along with the nucleic acid binding reagents. The control is used to identify sample-specific amplification inhibition, reducing the likelihood of false negatives and providing confidence that results are accurate.
Specifications
Specifications
| Concentration | 10,000 Copies/μL |
| Content And Storage | 5 tubes containing 200 μL Store at -15° to -25°C. |
| Detection Method | Primer-probe |
| Format | Tube |
| Product Line | TaqMan |
| Quantity | 5 x 200 μL |
| Shipping Condition | Dry ice |
| For Use With (Application) | Pathogen Detection |
| Volume | 1000 μL |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.