Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Vaginal Microbiota Amplification Control

Catalog No. A32913
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32913 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A32913 Supplier Applied Biosystems™ Supplier No. A32913
Add to Cart
Add to Cart
This item is not returnable. View return policy

The TaqMan™ Vaginal Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan Vaginal Microbiota assays. This plasmid serves as a positive control for the TaqMan Vaginal Microbiota Assay collection.

  • The TaqMan™ Vaginal Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan™ Vaginal Microbiota assays
  • It is a multi-target plasmid that includes amplicons for each of the qualified TaqMan Vaginal Microbiota assays, as well as control prokaryotic 16S rRNA and human RNase P RPPH1 gene sequences
  • This plasmid serves as a positive control for the TaqMan Vaginal Microbiota Assay collection
  • The TaqMan Vaginal Microbiota Amplification Control can be included in vaginal microbiota profiling experiments to verify assay performance and to assist with troubleshooting
TRUSTED_SUSTAINABILITY

Specifications

Concentration 105 Copies/μL
Content And Storage Store at -5 to -30°C.
GC-Rich PCR Performance Low
PCR Method qPCR
Label or Dye Unlabeled
Product Line TaqMan
Product Type Vaginal Microbiota Amplification Control
Quantity 250 μL
Shipping Condition Gel Packs

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.