Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqPath™ COVID-19, Flu A, Flu B, RSV Select Panel

Catalog No. A56123
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A56123 200 Reactions
A56124 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A56123 Supplier Applied Biosystems™ Supplier No. A56123
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A multiplex real-time PCR solution that contains the assay, master mix, and controls needed for the simultaneous detection of RNA from COVID-19, influenza A, influenza B, and RSV in extracted nasopharyngeal swab specimens.

The Applied Biosystems TaqPath COVID-19, Flu A, Flu B, RSV Select Panel is a multiplex real-time PCR solution that contains the assay, master mix, and controls needed for the detection and differentiation of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), influenza A, influenza B, and respiratory syncytial virus (RSV) in RNA extracted from nasopharyngeal (NP) swab and anterior nasal (AN) swab specimens. Human ribonuclease P (RNase P) is included as an endogenous sample collection control.

Features of the TaqPath COVID-19, Flu A, Flu B, RSV Select Panel include:

  • Single test for COVID-19, influenza A, influenza B, and RSV— detect and differentiate between infections with similar clinical signs and symptoms, with robust detection of co-infections
  • Multi-target assay design—multiple gene targets for SARS-CoV-2, flu A, flu B, and RSV to compensate for emerging mutations and variants and provide increased confidence in results
  • Complete integrated clinical solution—end-to-end sample to result workflow with laboratory information system (LIS) integration via Diomni Software

Each kit includes the following components:

  • TaqPath COVID‐19, Flu A, Flu B, RSV Select Assay—multiplex assay that contains primer and probe sets specific to the following targets:
    • Three SARS-CoV-2 targets (Orf1a, Orf1b, and N genes)
    • Two influenza A targets (PB1 & M genes)
    • Two influenza B targets (M & NS genes)
    • Three RSV targets (NP, M, and L protein genes)
    • RNase P (human sample collection control)
  • TaqPath COVID‐19, Flu A, Flu B, RSV Select Positive Control—inactivated viral control that contains SARS-CoV-2, influenza A, influenza B, and RSV
  • TaqPath COVID‐19, Flu A, Flu B, RSV Select Negative Control—MS2 packaged RNA control that contains targets specific to RNase P genomic regions targeted by the assay
  • TaqPath 1-Step Select Master Mix (No ROX)—ready-to-use PCR mix, including reverse transcriptase, polymerase, dNTPs, salt, and buffer
  • Package insert—provides the instructions and the link to download the instructions for use
  • Assay Definition File (ADF)—applicable to the instrument used in the workflow (available via download)

Intended use

The Applied Biosystems TaqPath COVID‐19, Flu A, Flu B, RSV Select Panel is a multiplex, real-time reverse transcription polymerase chain reaction (RT-PCR) in vitro diagnostic test for the qualitative detection and differentiation of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), influenza A virus, influenza B virus, and respiratory syncytial virus (RSV). Nucleic acids are isolated and purified from nasopharyngeal (NP) swab and anterior nasal (AN) swab specimens obtained from individuals exhibiting signs and symptoms of a respiratory tract infection. Clinical signs and symptoms of respiratory viral infection due to SARS-CoV-2, influenza, and RSV can be similar. This test is intended to aid in the differential diagnosis of SARS-CoV-2, influenza A, influenza B, and RSV A/B (undifferentiated) infections in humans and is not intended to detect influenza C virus infections.

Nucleic acids from the viral organisms identified by this test are generally detectable in NP and AN swab specimens during the acute phase of infection. The detection and identification of specific viral nucleic acids from individuals exhibiting signs and symptoms of respiratory tract infection are indicative of the presence of the identified virus and aids in diagnosis if used in conjunction with other clinical and epidemiological information, and laboratory findings.

The results of this test should not be used as the sole basis for diagnosis, treatment, or other patient management decisions. Positive results do not rule out coinfection with other organisms. The organism(s) detected by the Applied Biosystems TaqPath COVID‐19, Flu A, Flu B, RSV Select Panel may not be the definite cause of disease. Negative results do not preclude SARS-CoV-2, influenza A virus, influenza B virus, or RSV infections.

The Applied Biosystems TaqPath COVID‐19, Flu A, Flu B, RSV Select Panel is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of real-time PCR and in vitro diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Primer-probe
Form Solution
PCR Method RT-PCR
For Use With (Equipment) KingFisher™ Apex Dx 96 Deep-Well, QuantStudio™ 5 Dx Real‐Time PCR Instrument (96-well, 0.2-mL block)
Label or Dye ABY, FAM, JUN, VIC, Alexa Fluor 647
Quantity 200 Reactions
For Use With (Application) Pathogen Detection
No. of Reactions 200 Reactions
Can I use Diomni Software v4.2 with the TaqPath COVID-19, Flu A, Flu B, RSV Select Panel IVD test (Cat. No. A56123, A56124)?

No, at this time, only Diomni Software v4.1 is approved for use with the TaqPath COVID-19, Flu A, Flu B, RSV Select Panel IVD test.

For In Vitro Diagnostic Use. Not available in all countries or regions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.