Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tara Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551998 Supplier Invitrogen™ Supplier No. PA551998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EIGALMRQAE EREHTLRRCQ QEGQELLRHN QELHGRLSEE IDQLRGFIAS QGMGNGCGRS NERSSCELEV LLRVKENELQ YLKKEVQCLR DELQMMQKDK RFTSGKYQDV YVELSHIKTR SEREIEQLKE HLRL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

This gene encodes a protein with an N-terminal pleckstrin homology domain and a C-terminal coiled-coil region. The protein interacts with trio, which is involved with neural tissue development and controlling actin cytoskeleton organization, cell motility and cell growth. The protein also associates with F-actin and stabilizes F-actin structures. Mutations in this gene have been associated with a form of autosomal recessive nonsyndromic deafness. Multiple alternatively spliced transcript variants that would encode different isoforms have been found for this gene, however some transcripts may be subject to nonsense-mediated decay (NMD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tara
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIOBP
Gene Accession No. Q9H2D6
Gene Alias AI428493; DFNB28; dJ37E16.4; EST478828; HRIHFB2122; Kiaa1662; mKIAA1662; protein Tara; TAP68; TARA; tara-like protein; TRIO and F-actin binding protein; TRIO and F-actin-binding protein; Trio-associated repeat on actin; TRIOBP
Gene Symbols TRIOBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Tara. Recombinant protein control fragment (Product #RP-91495).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11078
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.