Learn More
TARC Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$319.00
Specifications
Antigen | TARC |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P97100UG
![]() |
Invitrogen™
500P97100UG |
100 μg |
Each of 1 for $319.00
|
|
|||||
Description
AA Sequence of recombinant protein: ARGTNVGREC CLEYFKGAIP LRKLKTWYQT SEDCSRDAIV FVTVQGRAIC SDPNNKRVKN AVKYLQSLER S. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TARC (CCL17). Anti-Human TARC (CCL17)-specific antibody was purified by affinity chromatography employing an immobilized Human TARC (CCL17) matrix. Sandwich ELISA: To detect Human TARC (CCL17) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TARC (CCL17) (500-P97Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TARC (CCL17). Western Blot: To detect hTARC by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hTARC is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.Specifications
TARC | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q92583 | |
6361 | |
E.coli-derived Recombinant Human TARC (CCL17). | |
Antigen affinity chromatography | |
CCL17 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
A-152E5.3; ABCD-2; CC chemokine TARC; C-C motif chemokine; C-C motif chemokine 17; C-C motif chemokine ligand 17; CCL17; chemokine (C-C motif) ligand 17; SCYA17; Scya17l; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; small-inducible cytokine A17; T cell-directed CC chemokine; TARC; Thymus and activation-regulated chemokine | |
CCL17 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.