Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TARP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570504
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570504 Supplier Invitrogen™ Supplier No. PA570504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Horse: 86%; Human: 100%; Mouse: 86%; Pig: 79%; Rat: 79%.

In some non-lymphoid tissues, the unrearranged T cell receptor gamma locus is expressed. The resulting transcript contains a subset of theT cell receptor gamma locus gene segments and is shorter than T cell receptor gamma transcripts expressed in lymphoid tissues. The downstream ORF is in the same frame as T cell receptor gamma and its protein product is similar to T cell receptor gamma locus proteins. The upstream ORF uses a different reading frame and encodes a novel protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TARP
Gene Accession No. 0
Gene Alias antigen, T-cell receptor; CD3G; T cell receptor gamma chain variable; TARP; T-cell receptor gamma-chain constant region; TCR gamma alternate reading frame protein; TCR- gamma V-IV; TCR V gamma 3J>gamma>2C>gamma>2; TCR V3-JP2-Cgamma2; TCRG; TCRGC1; TCRGC2; TCRGV; Vgamma9ChainTCRMOP
Gene Symbols TARP
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human TARP.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445347
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.