Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542862 Supplier Invitrogen™ Supplier No. PA542862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PEYIYTRLEM YNILKAEHDS ILAEKAEKDS KPIKVTLPDG KQVDAESWKT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 91%; Zebrafish: 85%.

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Threonyl-tRNA synthetase belongs to the class-II aminoacyl-tRNA synthetase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARS
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TARS1
Gene Accession No. P26639
Gene Alias D15Wsu59e; hypothetical protein LOC510075; MGC9344; TARS; TARS1; threonine tRNA ligase 1, cytoplasmic; threonine--tRNA ligase; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonine--tRNA ligase, cytoplasmic; Threonyl-tRNA synthetase; threonyl-tRNA synthetase 1; threonyl-tRNA synthetase, cytoplasmic; ThrRS; wu:fb07c05; wu:fb39c12; zgc:92586; zgc:92586 protein
Gene Symbols TARS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TARS (aa 51-100).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6897
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.