Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577090 Supplier Invitrogen™ Supplier No. PA577090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Threonyl-tRNA synthetase belongs to the class-II aminoacyl-tRNA synthetase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TARS1
Gene Accession No. P26639, Q5XHY5, Q9D0R2
Gene Alias D15Wsu59e; hypothetical protein LOC510075; MGC9344; TARS; TARS1; threonine tRNA ligase 1, cytoplasmic; threonine--tRNA ligase; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonine--tRNA ligase, cytoplasmic; Threonyl-tRNA synthetase; threonyl-tRNA synthetase 1; threonyl-tRNA synthetase, cytoplasmic; ThrRS; wu:fb07c05; wu:fb39c12; zgc:92586; zgc:92586 protein
Gene Symbols TARS1
Host Species Rabbit
Immunogen Recombinant full length Human TARS.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110960, 294810, 6897
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.