Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588808 Supplier Invitrogen™ Supplier No. PA588808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNGEPYDLER PLETDSDLRF LTFDSPEGKA VFWHSSTHVL GAAAEQFLGA VLCRGPSTEY GFYHDFFLGK ERTIRGSELP VLERICQELT AAARPFRRLE ASRDQLRQLF KDNPFKLHLI EEKVTGPTAT VYGCGTLVDL CQGPHLRHTG QIGGLKLLSN SSSLWRSSGA PETLQRVSGI SFPTTELLRV WEAWREEAEL RDHRRIGKEQ ELFFFHELSP GSCFFLPRGT RVYNALVAFI RAEYAHRGFS EVKTPTLFST KLWEQSGHWE HYQEDMFAVQ PPGSDRPPSS QSDDSTRHIT D; Positive Samples: LO2, U-87MG, Mouse brain; Cellular Location: Mitochondrion matrix.

This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.70 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TARS2
Gene Accession No. Q3UQ84, Q9BW92
Gene Alias 2610024N01Rik; AI429208; COXPD21; RGD1308283; TARS2; Tarsl1; threonine--tRNA ligase, mitochondrial; Threonyl-tRNA synthetase; threonyl-tRNA synthetase 2, mitochondrial (putative); threonyl-tRNA synthetase, mitochondrial; threonyl-tRNA synthetase-like 1; ThrRS
Gene Symbols TARS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human TARS2 (NP_0794262).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71807, 80222
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.