Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144027
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144027 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144027 Supplier Invitrogen™ Supplier No. PA5144027
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human Hela whole cell, human THP-1 whole cell, human Raji whole cell, rat liver tissue, mouse liver tissue. IHC: human colonic adenocarcinoma tissue, human lung cancer tissue, rat brain tissue, rat brain tissue. ICC/IF: T-47D cell. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARS2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene TARS2
Gene Accession No. Q3UQ84, Q68FW7, Q9BW92
Gene Alias 2610024N01Rik; AI429208; COXPD21; RGD1308283; TARS2; Tarsl1; threonine--tRNA ligase, mitochondrial; Threonyl-tRNA synthetase; threonyl-tRNA synthetase 2, mitochondrial (putative); threonyl-tRNA synthetase, mitochondrial; threonyl-tRNA synthetase-like 1; ThrRS
Gene Symbols TARS2
Host Species Rabbit
Immunogen E.coli-derived human TARS2 recombinant protein (Position: Q69-F718).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310672, 71807, 80222
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.