Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TARSH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169394
Description
TARSH Polyclonal specifically detects TARSH in Human samples. It is validated for Western Blot.Specifications
TARSH | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ABI family, member 3 (NESH) binding protein, ABI gene family member 3-binding protein, DKFZP586L2024, FLJ41743, Nesh-binding protein, NeshBP, NESHBPABI gene family, member 3 (NESH) binding protein, target of Nesh-SH3, Tarsh, TARSHFLJ41754 | |
Rabbit | |
119 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q7Z7G0 | |
ABI3BP | |
Synthetic peptides corresponding to TARSH or ABI3BP(ABI gene family, member 3 (NESH) binding protein) The peptide sequence was selected from the N terminal of TARSH. Peptide sequence LINPHHDWTLPSHCPNDRFYTIRYREKDKEKKWIFQICPATETIVENLKP. | |
Affinity purified | |
RUO | |
25890 | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction