Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TARSL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546633
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546633 Supplier Invitrogen™ Supplier No. PA546633
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TYVSKDGDDK KRPVIIHRAI LGSVERMIAI LSENYGGKWP FWLSPRQVMV Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Catalyzes the attachment of threonine to tRNA(Thr) in a two-step reaction: threonine is first activated by ATP to form Thr-AMP and then transferred to the acceptor end of tRNA(Thr). Also edits incorrectly charged tRNA(Thr) via its editing domain, at the post-transfer stage. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARSL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TARSL2
Gene Accession No. A2RTX5
Gene Alias A530046H20Rik; probable threonine--tRNA ligase 2, cytoplasmic; Probable threonyl-tRNA synthetase 2, cytoplasmic; RGD1311308; si:zfos-508h5.3; TARS3; TARSL2; threonine--tRNA ligase; Threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase 2, cytoplasmic; probable threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonyl-tRNA synthetase; threonyl-tRNA synthetase 3; threonyl-tRNA synthetase like 2; threonyl-tRNA synthetase-like 2; threonyl-tRNA synthetase-like protein 2; ThrRS
Gene Symbols TARSL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TARSL2 (aa 653-702).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123283
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.