Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TARSL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566861 Supplier Invitrogen™ Supplier No. PA566861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIPVGPTCEKY ALQVSSEFFE EGFMADVDLD HSCTL Highest antigen sequence identity to the following orthologs - mouse 86%, rat 58%.

Catalyzes the attachment of threonine to tRNA(Thr) in a two-step reaction: threonine is first activated by ATP to form Thr-AMP and then transferred to the acceptor end of tRNA(Thr). Also edits incorrectly charged tRNA(Thr) via its editing domain, at the post-transfer stage. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARSL2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TARSL2
Gene Accession No. A2RTX5
Gene Alias A530046H20Rik; probable threonine--tRNA ligase 2, cytoplasmic; Probable threonyl-tRNA synthetase 2, cytoplasmic; RGD1311308; si:zfos-508h5.3; TARS3; TARSL2; threonine--tRNA ligase; Threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase 2, cytoplasmic; probable threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonyl-tRNA synthetase; threonyl-tRNA synthetase 3; threonyl-tRNA synthetase like 2; threonyl-tRNA synthetase-like 2; threonyl-tRNA synthetase-like protein 2; ThrRS
Gene Symbols TARSL2
Host Species Rabbit
Immunogen Recombinant Human TARSL2. Recombinant protein control fragment (Product #RP-107514).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123283
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.