Learn More
Invitrogen™ TARSL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120406
Description
Positive test controls include: U-87MG, A-549, LO2, Mouse brain, Mouse heart, Mouse liver, Rat pancreas. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MAAEALAAEA VASRLERQEE DIRWLWSEVE RLRDEQLNAP YSCQAEGPCL TREVAQLRAE NCDLRHRLCS LRLCLAEERS RQATLESAEL EAAQEAGAQP PPSQSQDKDM KKKKMKESEA DSEVKHQPIF IKERLKLFEI LKKDHQLLLA IYGKKGDTSN IITVRVADGQ.
Catalyzes the attachment of threonine to tRNA(Thr) in a two-step reaction: threonine is first activated by ATP to form Thr-AMP and then transferred to the acceptor end of tRNA(Thr). Also edits incorrectly charged tRNA(Thr) via its editing domain, at the post-transfer stage. [UniProt].
Specifications
| TARSL2 | |
| Polyclonal | |
| Unconjugated | |
| TARSL2 | |
| A530046H20Rik; probable threonine--tRNA ligase 2, cytoplasmic; Probable threonyl-tRNA synthetase 2, cytoplasmic; RGD1311308; si:zfos-508h5.3; TARS3; TARSL2; threonine--tRNA ligase; Threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase 2, cytoplasmic; probable threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonyl-tRNA synthetase; threonyl-tRNA synthetase 3; threonyl-tRNA synthetase like 2; threonyl-tRNA synthetase-like 2; threonyl-tRNA synthetase-like protein 2; ThrRS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 123283, 272396, 308701 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.47 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| A2RTX5, Q8BLY2 | |
| TARSL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human TARSL2 (NP_689547.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.