Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tartrate Resistant Acid Phosphatase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584356
Description
Immunogen sequence: GGVPNAPFHT AREMANAKEI ARTVQILGAD FILSLGDNFY FTGVQDINDK RFQETFEDVF SDRSLRKVPW YVLAGNHDHL GNVSAQI.
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.
Specifications
Tartrate Resistant Acid Phosphatase | |
Polyclonal | |
Unconjugated | |
ACP5 | |
acid phosphatase 5, tartrate resistant; acid phosphatase type V; Acp5; HPAP; human purple acid phosphatase; T5ap; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; tartrate-resistant acid phosphatase type 5; TRACP; TRACP5a; TRACP5b; TRAP; TR-AP; TRAPase; TrATPase; TTRRAP; Type 5 acid phosphatase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P13686 | |
ACP5 | |
Recombinant protein corresponding to Human ACP5. Recombinant protein control fragment (Product #RP-103307). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction