Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tartrazine
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It can be employed as a biological stain and a food colorant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be employed as a biological stain and a food colorant.
Solubility
Soluble in water 260 g/L (30°C).
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place.
It can be employed as a biological stain and a food colorant.
Solubility
Soluble in water 260 g/L (30°C).
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place.

Specifications
Specifications
Quantity | 500 g |
Sensitivity | Hygroscopic |
Merck Index | 14,9072 |
Solubility Information | Soluble in water 260g/L (30°C). |
Formula Weight | 534.37 |
Chemical Name or Material | Tartrazine |
Safety and Handling
Hazard Category | H317-H334 |
Hazard Statement | GHS H Statement H334-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
Precautionary Statement | P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 217-699-5 |
RTECSNumber | UQ6400000 |
TSCA | Yes |
storageNote1 | Hygroscopic |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction