Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tartronic acid, 97%

Catalog No. AAB2164002
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2164002 0.5 g
AAB2164077 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2164002 Supplier Thermo Scientific Chemicals Supplier No. B2164002
Add to Cart

CAS: 80-69-3 | C3H4O5 | 120.06 g/mol

Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.

Solubility
Soluble in water and alcohol.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-69-3
Molecular Formula C3H4O5
Molecular Weight (g/mol) 120.06
MDL Number MFCD00004237
InChI Key ROBFUDYVXSDBQM-UHFFFAOYSA-N
Synonym tartronic acid, hydroxymalonic acid, 2-hydroxymalonic acid, propanedioic acid, hydroxy, tartronicacid, 2-tartronic acid, hydroxypropanedioic acid, 2-hydroxymalonate, malonic acid, hydroxy, hydroxymalonate
PubChem CID 45
ChEBI CHEBI:16513
IUPAC Name 2-hydroxypropanedioic acid
SMILES C(C(=O)O)(C(=O)O)O

Specifications

Melting Point ∼151°C (decomposition)
Assay Percent Range 0.98
Beilstein 1209791
Merck Index 14,9073
Solubility Information Soluble in water and alcohol.
Formula Weight 120.06
Percent Purity 98%
Quantity 0.5 g
Chemical Name or Material Tartronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-301-1
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.