Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAS1R1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132846
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132846 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132846 Supplier Thermo Scientific Supplier No. PA132846

Rabbit Polyclonal Antibody

PA1-32846 detects GPCR TAS1R1 from human samples.

Putative taste receptor. TAS1R1/TAS1R3 responds to the umami taste stimulus (the taste of monosodium glutamate) and also to most of the 20 standard L-amino acids, but not to their D-enantiomers or other compounds. Sequence differences within and between species can significantly influence the selectivity and specificity of taste responses. Expressed strongly only in fungiform papillae.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS1R1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TAS1R1
Gene Accession No. Q7RTX1
Gene Alias candidate taste receptor T1R1; G protein-coupled receptor 70; GM148; GPCRB3; GPCR-B3; GPR70; G-protein coupled receptor 70; seven transmembrane helix receptor; T1r1; TAS1R1; taste 1 receptor member 1; taste receptor type 1 member 1; taste receptor, type 1, member 1; TR1; TS1R1
Gene Symbols TAS1R1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human TAS1R1
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80835
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.