Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS1R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116736 Supplier Invitrogen™ Supplier No. PA5116736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, HeLa, 293T Immunogen sequence: IVQWQWDRSQ NPFQSVASYY PLQRQLKNIQ DISWHTINNT IPMSMCSKRC QSGQKKKPVG IHVCCFECID CLPGTFLNHT EDEYECQACP NNEWSYQSET S.

Putative taste receptor. TAS1R2/TAS1R3 recognizes diverse natural and synthetic sweeteners. Expressed mainly in circumvallate and foliate taste papillae.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS1R2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TAS1R2
Gene Accession No. Q8TE23
Gene Alias aldehyde dehydrogenase 4 family, member A1; Aldh4a1; candidate taste receptor T1R2; G protein-coupled receptor 71; GPCRB4; GPR71; G-protein coupled receptor 71; Sweet taste receptor T1R2; T1R2; Tas1r2; taste 1 receptor member 2; taste receptor TR2; taste receptor type 1 member 2; taste receptor, type 1, member 2; Tr2; TS1R2
Gene Symbols TAS1R2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 450-550 of human TAS1R2 (NP_6894182).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80834
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.