Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS1R3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA132795

Catalog No. PIPA132795


Only null left
Add to Cart

Description

Description

PA1-32795 detects GPCR TAS1R3 from human samples.

The protein encoded by T1R3 is a G-protein coupled receptor involved in taste responses. The encoded protein can form a heterodimeric receptor with TAS1R1 to elicit the umami taste response, or it can bind with TAS1R2 to form a receptor for the sweet taste response.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TAS1R3
Polyclonal
Unconjugated
TAS1R3
Sac; Saccharin preference protein; sweet taste receptor T1R3; T1r3; TAS1R3; taste 1 receptor member 3; taste receptor type 1 member 3; taste receptor, type 1, member 3; TR3; TS1R3
Rabbit
Antigen affinity chromatography
RUO
83756
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q7RTX0
TAS1R3
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human T1R3.
25 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.