Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAS1R3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102260 Supplier Thermo Scientific Supplier No. PA5102260
Add to Cart
Add to Cart

TAS1R3 Polyclonal Antibody

TAS1R3 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS1R3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAS1R3
Gene Accession No. Q7RTX0
Gene Alias Sac; Saccharin preference protein; sweet taste receptor T1R3; T1r3; TAS1R3; taste 1 receptor member 3; taste receptor type 1 member 3; taste receptor, type 1, member 3; TR3; TS1R3
Gene Symbols TAS1R3
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAS1R3(Accession Q7RTX0), corresponding to amino acid residues E333-A383.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83756
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.