Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS1R3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534255
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534255 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534255 Supplier Invitrogen™ Supplier No. PA534255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Baboon (100%) Orangutan (89%).

The protein encoded by T1R3 is a G-protein coupled receptor involved in taste responses. The encoded protein can form a heterodimeric receptor with TAS1R1 to elicit the umami taste response, or it can bind with TAS1R2 to form a receptor for the sweet taste response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS1R3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TAS1R3
Gene Accession No. Q7RTX0
Gene Alias Sac; Saccharin preference protein; sweet taste receptor T1R3; T1r3; TAS1R3; taste 1 receptor member 3; taste receptor type 1 member 3; taste receptor, type 1, member 3; TR3; TS1R3
Gene Symbols TAS1R3
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human TAS1R3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83756
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.