Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144983 Supplier Invitrogen™ Supplier No. PA5144983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAYIAKILND YKMKNDTVWD LNMYK Antigen sequence identity: Mouse 40%; Rat 40%.

This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAS2R10
Gene Accession No. Q9NYW0
Gene Alias MGC126811; MGC126813; T2R10; TAS2R10; taste 2 receptor member 10; taste receptor family B member 2; Taste receptor type 2 member 10; taste receptor, family B, member 2; taste receptor, type 2, member 10; TRB2
Gene Symbols TAS2R10
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 144-168. Recombinant protein control fragment (Product #RP-109852).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.