Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAS2R10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590261
Description
Immunogen sequence: TSLSIFYFLK IANFSNYIFL WLKSRTNMVL PFMIVFLLIS SLLNFAYIAK ILNDYKTKND TVWDLNMYKS EYFIKQILLN LGVIFFFTLS LITCIFLIIS L; Positive Samples: Mouse brain, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.
This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13.
Specifications
TAS2R10 | |
Polyclonal | |
Unconjugated | |
Tas2r110 | |
MGC126811; MGC126813; T2R10; TAS2R10; taste 2 receptor member 10; taste receptor family B member 2; Taste receptor type 2 member 10; taste receptor, family B, member 2; taste receptor, type 2, member 10; TRB2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100310875, 387344 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.12 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q7M712, Q9JKE8 | |
Tas2r110 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAS2R10 (NP_0764101). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction