Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAS2R14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP280531
Description
TAS2R14 Polyclonal specifically detects TAS2R14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TAS2R14 | |
Polyclonal | |
Western Blot 1:300 - 1:2000, Immunohistochemistry 1:300 - 1:2000, Immunohistochemistry-Paraffin 1:300 - 1:2000 | |
MGC125491, T2R14MGC125492, Taste receptor family B member 1, taste receptor, family B, member 1, taste receptor, type 2, member 14, TRB1taste receptor type 2 member 14 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Lyophilized from whole serum | |
TAS2R14 | |
A synthetic peptide from amino acid region 250-317 of human TAS2R14 conjugated to blue carrier protein was used as the antigen. | |
0.1 mL | |
GPCR, Protein Kinase | |
50840 | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction