Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102253 Supplier Invitrogen™ Supplier No. PA5102253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAS2R48.

Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R19
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAS2R19
Gene Accession No. P59542
Gene Alias MGC126853; MGC138203; MSTP058; T2R19; T2R23; T2R48; TAS2R19; TAS2R23; TAS2R48; taste 2 receptor member 19; taste receptor type 2 member 19; Taste receptor type 2 member 23; Taste receptor type 2 member 48; taste receptor, type 2, member 19; taste receptor, type 2, member 23; taste receptor, type 2, member 48
Gene Symbols TAS2R19
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAS2R19(Accession P59542), corresponding to amino acid residues L185-L235.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259294
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.