Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599957 Supplier Invitrogen™ Supplier No. PA599957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAS2R47.

Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R30
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAS2R30
Gene Accession No. P59541
Gene Alias T2R30; T2R44; T2R47; T2R53; TAS2R30; TAS2R44; TAS2R47; taste 2 receptor member 30; taste receptor T2R47; taste receptor type 2 member 30; Taste receptor type 2 member 47; taste receptor, type 2, member 30; taste receptor, type 2, member 47; type 2 taste receptor member 30; type 2 taste receptor member 47
Gene Symbols TAS2R30
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAS2R30(Accession P59541), corresponding to amino acid residues C238-Q288.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259293
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.