Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560539 Supplier Invitrogen™ Supplier No. PA560539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLGRHMRTMK VYTRNSRDPS LEAHIKALKS Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 70%.

Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Variations in TAS2R38 are associated with the ability to taste phenylthiocarbamide (PTC tasting); also called thiourea tasting. The ability to taste the substance PTC and a number of related substances is genetically controlled. Genetic studies have demonstrated complex inheritance for this trait. For some people (and some chimpanzees also), the chemical PTC tastes very bitter. For others, it is tasteless. Actually, substantial variation in taste sensitivity exists in human. Five haplotypes arising from three coding SNPs in the TAS2R38 gene are associated with distinct phenotypes of PTC taste sensitivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R38
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAS2R38
Gene Accession No. P59533
Gene Alias mt2r31; PTC; PTC bitter taste receptor; RGD:1560649}; T2R138; T2R26; T2r31; T2r38; T2r38 {ECO:0000312; T2R61; Tas2r138; Tas2r26; TAS2R38; taste 2 receptor member 38; taste receptor type 2 member 26; Taste receptor type 2 member 38; taste receptor type 2 member 61; taste receptor, type 2, member 138; taste receptor, type 2, member 38
Gene Symbols TAS2R38
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAS2R38. Recombinant protein control fragment (Product #RP-95331).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5726
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.