Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAS2R38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562953
Description
Immunogen sequence: NAKLRRAVMT ILLWAQSSLK VRADHKADSR TL Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 53%.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Variations in TAS2R38 are associated with the ability to taste phenylthiocarbamide (PTC tasting); also called thiourea tasting. The ability to taste the substance PTC and a number of related substances is genetically controlled. Genetic studies have demonstrated complex inheritance for this trait. For some people (and some chimpanzees also), the chemical PTC tastes very bitter. For others, it is tasteless. Actually, substantial variation in taste sensitivity exists in human. Five haplotypes arising from three coding SNPs in the TAS2R38 gene are associated with distinct phenotypes of PTC taste sensitivity.
Specifications
| TAS2R38 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R38 | |
| mt2r31; PTC; PTC bitter taste receptor; RGD:1560649}; T2R138; T2R26; T2r31; T2r38; T2r38 {ECO:0000312; T2R61; Tas2r138; Tas2r26; TAS2R38; taste 2 receptor member 38; taste receptor type 2 member 26; Taste receptor type 2 member 38; taste receptor type 2 member 61; taste receptor, type 2, member 138; taste receptor, type 2, member 38 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P59533 | |
| TAS2R38 | |
| Recombinant protein corresponding to Human TAS2R38. Recombinant protein control fragment (Product #RP-95332). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction