Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R39 Polyclonal Antibody

Catalog No. PIPA563888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563888 Supplier Invitrogen™ Supplier No. PA563888

Rabbit Polyclonal Antibody

Immunogen sequence: ICTVYCNNSF PIHSSNSTKK TYLSEINVVG L Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 52%.

The protein encoded by this gene is a bitter taste receptor that detects green tea catechins, soy isoflavones, and theaflavins. The encoded protein is gustducin-linked and may activate alpha gustducin. This gene is intronless.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R39
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAS2R39
Gene Accession No. P59534
Gene Alias T2R39; T2R57; TAS2R39; taste 2 receptor member 39; taste receptor type 2 member 39; Taste receptor type 2 member 57; taste receptor, type 2, member 39
Gene Symbols TAS2R39
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAS2R39. Recombinant protein control fragment (Product #RP-100571).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259285
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.