Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAS2R4 Antibody, Novus Biologicals™
SDP

Catalog No. NB018013 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018013 0.025 mL
NB11074890 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018013 Supplier Novus Biologicals Supplier No. NB110748900.025ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAS2R4 Polyclonal specifically detects TAS2R4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen TAS2R4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000
Formulation Whole antisera with No Preservative
Gene Accession No. Q9NYW5
Gene Alias candidate taste receptor T2R4, taste receptor, type 2, member 4
Gene Symbols TAS2R4
Host Species Rabbit
Immunogen A synthetic peptide as a part of human taste receptor, type 2, member 4 (TAS2R4, TA2R4, T2R4) conjugated to immunogenic carrier protein was used as the immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 50832
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.