Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599931 Supplier Invitrogen™ Supplier No. PA599931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAS2R4.

This gene encodes a member of a family of candidate taste receptors that are members of the G protein-coupled receptor superfamily and that are specifically expressed by taste receptor cells of the tongue and palate epithelia. These apparently intronless genes encode a 7-transmembrane receptor protein, functioning as a bitter taste receptor. This gene is clustered with another 3 candidate taste receptor genes in chromosome 7 and is genetically linked to loci that influence bitter perception. Gustducin-coupled receptor for denatonium and N(6)-propyl-2-thiouracil implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and palate epithelium and exclusively in gustducin-positive cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAS2R4
Gene Accession No. Q67ET0, Q9JKT3, Q9NYW5
Gene Alias bitter taste receptor T2R16; candidate taste receptor T2R4; candidate taste receptor T2R8; MGC163311; mGR08; mt2r8; T2R108; T2R16; T2R20; T2R4; T2R8; Tas2r108; TAS2R4; tas2r4 {ECO:0000250; Tas2r8; taste 2 receptor member 4; taste receptor T2R8; taste receptor type 2 member 20; Taste receptor type 2 member 4; taste receptor type 2 member 8; taste receptor, type 2, member 108; taste receptor, type 2, member 4; UniProtKB:Q9JKT3}
Gene Symbols Tas2r108, TAS2R4
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAS2R4(Accession Q9NYW5), corresponding to amino acid residues F49-V99.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50832, 554302, 57253
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.