Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599929 Supplier Invitrogen™ Supplier No. PA599929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAS2R40.

Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Several bitter taste receptors are expressed in a single taste receptor cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R40
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAS2R40
Gene Accession No. P59535, Q67ET2, Q7TQB8
Gene Alias G protein-coupled receptor 60; GPR60; G-protein coupled receptor 60; MGC126844; MGC126846; mt2r33; RGD1562314; T2R18; T2r33; T2R40; T2R58; Tas2r144; TAS2R40; tas2r40 {ECO:0000250; Tas2r44; taste 2 receptor member 40; taste receptor type 2 member 18; Taste receptor type 2 member 40; taste receptor type 2 member 58; taste receptor, type 2, member 144; taste receptor, type 2, member 40; UniProtKB:P59535}
Gene Symbols Tas2r144, TAS2R40
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAS2R40(Accession P59535), corresponding to amino acid residues F156-L206.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259286, 387515, 500101
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.