Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAS2R41 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23381325UL
Description
TAS2R41 Polyclonal specifically detects TAS2R41 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TAS2R41 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
P59536 | |
TAS2R41 | |
This antibody was developed against a recombinant protein corresponding to amino acids: NFYYSAQKVEYSGGLGRQFFH | |
25 μL | |
GPCR | |
259287 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MGC119928, T2R41, T2R59MGC119929, taste receptor type 2 member 41, Taste receptor type 2 member 59, taste receptor, type 2, member 41 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction