Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144106
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144106 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144106 Supplier Invitrogen™ Supplier No. PA5144106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAS2R41 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

TAS2R41 is a receptor that may play a role in the perception of bitterness and belongs to the G-protein receptor superfamily. TAS2R41 is expressed mainly in taste receptor cells of the tongue and the palate eptithelia. Structurally, the TAS2R41 gene encodes a seven trans-membrane receptor protein and is clustered together with eight other taste receptor genes on chromosome 7. The compound chloramphenicol is an agonist for the TAS2R41 protein. Further, TAS2R41 may play a role in sensing the chemical composition of the gastrointestinal content. The activity of TAS2R41 may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. TAS2R41 is expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R41
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TAS2R41
Gene Accession No. P59536
Gene Alias MGC119928; MGC119929; mGR26; mt2r35; T2R12; T2R26; T2R41; T2R59; Tas2r12; Tas2r126; Tas2r26; Tas2r41; taste 2 receptor member 41; taste receptor rT2R12; taste receptor type 2 member 41; Taste receptor type 2 member 59; taste receptor, type 2, member 126; taste receptor, type 2, member 41
Gene Symbols TAS2R12P, TAS2R41
Host Species Rabbit
Immunogen Recombinant Human Taste receptor type 2 member 41 protein (205-234AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259287, 266656
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.