Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAS2R42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557544 Supplier Invitrogen™ Supplier No. PA557544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLVRHTRNLK LSSLGSRDSS TEAHRRAMK Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 62%.

Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R42
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAS2R42
Gene Accession No. Q7RTR8
Gene Alias candidate taste receptor hT2R55; hT2R55; T2R24; T2R42; T2R55; TAS2R42; TAS2R55; taste 2 receptor member 42; taste receptor type 2 member 42; taste receptor type 2 member 55; taste receptor, type 2, member 42
Gene Symbols TAS2R42
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAS2R42. Recombinant protein control fragment (Product #RP-95168).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353164
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.