Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAS2R43 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TAS2R43 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TAS2R43 Polyclonal specifically detects TAS2R43 in Human samples. It is validated for Western Blot.Specifications
TAS2R43 | |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS buffer, 2% sucrose | |
259289 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
T2R43, T2R52Taste receptor type 2 member 52, taste receptor type 2 member 43, taste receptor, type 2, member 43 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of human TAS2R43 (NP_795365.2). Peptide sequence MLANLVPFTVTLISFLLLVCSLCKHLKKMQLHGKGSQDPSTKVHIKVLQT | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title