Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAS2R45 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5102300
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102300 Supplier Thermo Scientific Supplier No. PA5102300

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAS2R45.

Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R45
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAS2R45
Gene Accession No. P59539
Gene Alias GPR59; G-protein coupled receptor 59; MGC119923; MGC119924; MGC119925; T2R45; TAS2R45; taste 2 receptor member 45; taste receptor type 2 member 45; taste receptor, type 2, member 45; ZG24P
Gene Symbols TAS2R45
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAS2R45(Accession P59539), corresponding to amino acid residues Q229-L279.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259291
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.